Order Entry
Germany
ContactUsLinkComponent
Sodium hydroxide
Sodium hydroxide
Catalog # VWRS5814.9010
Supplier:  VWR Collection
CAS Number:  
Sodium hydroxide
Catalog # VWRS5814.9010
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    10 kg
  • Seq:
    4158

Specifications

About this item

CAS Number:1310-73-2
 :
Formula:NaOH
MDL Number:MFCD00003548
 :40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
 :GHS05