Order Entry
Germany
ContactUsLinkComponent
Sodium hydroxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Corrosive
Sodium hydroxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0403.5000
Supplier:  Avantor
CAS Number:  
Corrosive
Sodium hydroxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0403.5000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    min. 99.0 %
  • Ammonium Hydroxide Precipitate:
    max. 0.02 %
  • Chloride (Cl):
    max. 5 ppm
  • Potassium (K):
    max. 0.01 %
  • Heavy Metals (as Ag):
    max. 5 ppm
  • Heavy Metals (as Pb):
    max. 5 ppm
  • Na2CO3:
    max. 1.0 %
  • Iron (Fe):
    max. 3
  • Nickel (Ni):
    max. 5
  • Mercury (Hg) (by AAS):
    max. 0.1
  • Nitrogen Compounds (as N):
    max. 3
  • Phosphate (as PO4):
    max. 10
  • Sulfate (SO4):
    max. 5

Specification Test Results

About this item

 :
 :40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548