Sodium peroxide ≥93%, granules, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1313-60-6
:
Formula:Na₂O₂
MDL Number:MFCD00003497
:77.98 g/mol
Merck Index:13,08728
Melting Pt:660 °C
Density:2.805 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-209-4
UN:1504
ADR:5.1,I
:GHS03
:GHS05
Specification Test Results
- Assay:Min. 93 %
- Identification:Passes test
- Mean diameter:0.3 to 2.0 mm
- Heavy metals (as Pb):Max. 20 ppm
- Total N (Nitrogen):Max. 20 ppm
- Cl (Chloride):Max. 20 ppm
- PO4 (Phosphate):Max. 5 ppm
- SiO2 + Al2O3 + Fe2O3:Max. 0.025 %
- SO4 (Sulphate):Max. 50 ppm
- Al (Aluminium):Max. 10 ppm
- Ca (Calcium):Max. 0.05 %
- Fe (Iron):Max. 20 ppm