Order Entry
Germany
ContactUsLinkComponent
Sodium peroxide 96%
Sodium peroxide 96%
Certificates
undefined
Sodium peroxide 96%
Certificates

Specification Test Results

  • Appearance:
    Yellow Granules (May contain a small quantity of discolored material from the manufacturing process)
  • Titration with KMnO4:
    ≥95 %
  • Chloride (Cl):
    ≤50 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm

Specification Test Results

Related Information