Order Entry
Germany
Orders LinkContactUsLinkComponent
Water

Water

   Sustainable Options Available
Marketsource
Supplier:   Serva
Water
Supplier:  Serva
   Sustainable Options Available
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SERA38381.01
  • SERA38381.03
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL

    Specifications:
      Safer Human and Environmental Health
    Marketsource
    More
    Specifications
    Cat. No.
    SERA38381.01
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
      Safer Human and Environmental Health
    Marketsource
    More
    Specifications
    Cat. No.
    SERA38381.03
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.