Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Water

   Sustainable Options Available
Supplier:   Bernd Kraft
Water
Supplier:  Bernd Kraft
   Sustainable Options Available
Certificates

5 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • KRAF01870.4000
  • KRAF01870.3000
  • KRAF01870.5000
  • KRAF01870.6000
  • KRAF01870.7020
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Environmental certification:
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 28,40 €
    28,40 €
    1 * 2.500 mL
       28,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF01870.4000
    Environmentally Preferable
    Environmental certification
  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 19,40 €
    19,40 €
    1 * 1 L
       19,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF01870.3000
    Environmentally Preferable
    Environmental certification
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 46,90 €
    46,90 €
    1 * 5 L
       46,90 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF01870.5000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 76,20 €
    76,20 €
    1 * 10 L
       76,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF01870.6000
    Environmentally Preferable
    Environmental certification
  • Plastic bottle
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L

    Specifications:
    More
    Specifications
    Cat. No.
    KRAF01870.7020
    Environmentally Preferable
    Environmental certification