Order Entry
Germany
ContactUsLinkComponent
Water, HyClone™ HyPure, Cell Culture Grade
Water, HyClone™ HyPure, Cell Culture Grade
Certificates
undefined
Water, HyClone™ HyPure, Cell Culture Grade
Certificates

Specification Test Results

  • Endotoxin Concentration:
    < 0,005 EU/mL
  • Sterilisation:
    0,1 µm sterile filtered
  • pH:
    5,0 to 7,0
  • Storage:
    2° to 30° C

Specification Test Results

Related Information