Socket GL tube fittings, BOLA
Supplier: Bohlender
Certificates
About this item
PTFE, white.
- Temperature resistance: −200 to +250 °C
- Universal chemical resistance
- Vacuum: suitable
Fitting with socket for tubes with ground joint and GL thread for connection of hard-walled tubing (e.g. PTFE, PFA, FEP) with laboratory screw joints. The products is only exposed to PTFE.