Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Water, deionised

Supplier:   Hach
Water, deionised
Supplier:  Hach
Certificates
Water, deionised

2 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2

 

Product Family Options

Product Information