Order Entry
Germany
Orders LinkContactUsLinkComponent
Water, Proteomics Grade

Water, Proteomics Grade

Glass bottle, 1 L
Catalog # 786-229
Supplier:   G-Biosciences
Water, Proteomics Grade
Catalog # 786-229
Supplier:  G-Biosciences
CAS Number:  7732-18-5
Water, Proteomics Grade

Glass bottle, 1 L

CAS Number:  7732-18-5
Price
51,50 €
Log in for availability
1 * 1 L
Product Details & Documents

Ultra pure proteomic grade water that is fully compatible with 2D electrophoresis and mass spectrometry analysis.

Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.