Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium nitrate ≥98%

Sodium nitrate ≥98%

Oxidizing
Irritant
Sodium nitrate ≥98%
Certificates
Sodium nitrate ≥98%
Sodium nitrate ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Odour: Odourless Melting Point: ca 308° UN No.: 1498 MDL No.: MFCD00011119

  • CAS Number:7631-99-4
  • Formula:NaNO₃
  • MDL Number:MFCD00011119
  • Molecular Weight:84.99 g/mol
  • Merck Index:14,8647
  • Boiling Pt:380 °C (1013 hPa)
  • Melting Pt:∼308 °C
  • Density:2.26
  • Storage Temperature:Ambient
  • EINECS:231-554-3
  • UN:1498
  • ADR:5.1,III
  • REACH:01-2119488221-41
Product Family Options

Product Information