Sodium nitrite ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 7632-00-0 Molecular Formula: NNaO2 Formula Weight: 69.00 Odour: Odourless Melting Point: 271° dec. UN No.: 1500 MDL No.: MFCD00011118
Synonyms: Diazotizing salts
- CAS Number:7632-00-0
- Formula:NaNO₂
- MDL Number:MFCD00011118
- Molecular Weight:69 g/mol
- Merck Index:14,8648
- Boiling Pt:320 °C (1013 hPa)
- Melting Pt:271 °C (dec.)
- Density:2.17
- Storage Temperature:Ambient
- EINECS:231-555-9
- UN:1500
- ADR:5.1,III
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 7632-00-0 Molecular Formula: NNaO2 Formula Weight: 69.00 Odour: Odourless Melting Point: 271° dec. UN No.: 1500 MDL No.: MFCD00011118
Synonyms: Diazotizing salts
- CAS Number:7632-00-0
- Formula:NaNO₂
- MDL Number:MFCD00011118
- Molecular Weight:69 g/mol
- Merck Index:14,8648
- Boiling Pt:320 °C (1013 hPa)
- Melting Pt:271 °C (dec.)
- Density:2.17
- Storage Temperature:Ambient
- EINECS:231-555-9
- UN:1500
- ADR:5.1,III



