Order Entry
Germany
ContactUsLinkComponent
Potassium permanganate ≥99.0% (by manganometry), EMSURE® ACS for analysis (max. 0.05 ppm Hg), Supelco®
Oxidizing
Corrosive
Health
Irritant
Environment
Potassium permanganate ≥99.0% (by manganometry), EMSURE® ACS for analysis (max. 0.05 ppm Hg), Supelco®
Catalog # 1.05084.1000
Supplier:  Merck
CAS Number:  
Oxidizing
Corrosive
Health
Irritant
Environment
Potassium permanganate ≥99.0% (by manganometry), EMSURE® ACS for analysis (max. 0.05 ppm Hg), Supelco®
Catalog # 1.05084.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (manganometric):
    ≥ 99.0 %
  • Insoluble matter:
    ≤ 0.1 %
  • Chloride, Chlorate (as Cl):
    ≤ 0.005 %
  • Sulfate (SO₄):
    ≤ 0.01 %
  • Total nitrogen (N):
    ≤ 0.003 %
  • Cu (Copper):
    ≤ 0.001 %
  • Fe (Iron):
    ≤ 0.002 %
  • Hg (Mercury):
    ≤ 0.000005 %
  • Pb (Lead):
    ≤ 0.002 %

Specification Test Results

About this item

 :
 :158.03 g/mol
Merck Index:12,07824
Melting Pt:> 240 °C
Density:2.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-760-3
UN:1490
ADR:5.1,II
 :M-Clarity™ quality level = MQ300
 :GHS03
 :GHS05
 :GHS08
 :GHS07
 :GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364