Sodium periodate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:7790-28-5
:
:NaIO₄
:MFCD00003534
:13,08714
:300 °C (1013 hPa)
:175 °C
:3.87 g/cm³ (20 °C)
:232-197-6
:5.1,II
:213.89 g/mol
:1479
:GHS03
:Ambient
- Assay:Min. 99.0 %
- Assay (dried basis):99.8-100.3 %
- Cl + ClO3 + Br + BrO3 (as Cl):Max. 100 ppm
- SO4 (Sulphate):Max. 50 ppm
- Mn (Manganese):Max. 1 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test