


Potassium iodate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7758-05-6
:
Formula:KlO₃
MDL Number:MFCD00011406
:214 g/mol
Merck Index:13,07726
Melting Pt:560 °C
Density:3.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-831-9
ADR:5.1,II
:M-Clarity™ quality level = MQ300
UN:1479
:GHS03
:GHS05
:GHS07
Specification Test Results
- Solubility:92 g/l (25 °C)
- Melting Point:560 °C decomposes
- Molar Mass:214 g/mol
- Bulk Density:2000 kg/m3
- Density:3.98 g/cm3 (20 °C)
- pH:6 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Related Information
Frequently Bought Together