Potassium iodide ≥99% ACS
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Merck Index:13,07727
Boiling Pt:1330 °C (1013 hPa)
Melting Pt:681 °C
Density:3.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-659-4
:GHS07
:GHS08
Specification Test Results
- Barium:<0.002%
- Calcium:≤0.002%
- Chloride & Bromide:≤0.01%
- Heavy Metals (as Pb):≤0.0005%
- Insolubles:≤0.005%
- Iodate:≤0.0003%
- Iron:≤0.0003%
- Loss on Drying:≤0.2%
- Magnesium:≤0.001%
- pH (5%, Water) @25 °C:6 – 9.2
- Phosphate:≤0.001%
- Purity:≥99%
- Sodium:≤0.005%
- Sulphate:≤0.005%