Potassium iodide ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Merck Index:14,7643
Boiling Pt:1330 °C
Melting Pt:681 °C
Density:3.12
Storage Temperature:Ambient
EINECS:231-659-4