Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium iodide
Potassium iodide ≥99.99% (metals basis)
Potassium iodide ≥99.99% (metals basis)
:
Thermo Fisher Scientific
Potassium iodide ≥99.99% (metals basis)
:
Thermo Fisher Scientific
:
7681-11-0
:
:
Kl
:
MFCD00011405
:
166 g/mol
:
14,7643
:
1330 °C
:
681 °C
:
3.12
:
Ambient
:
213-659-4
msds_ALFA087627
Thermo Scientific Acroseal 25 Years