Potassium iodide 99%, briquettes, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Merck Index:13,07727
Boiling Pt:1330 °C (760 mmHg)
Melting Pt:680 °C
Density:3,13 g/cm³ (20 °C)
EINECS:231-659-4
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Briquettes
- Titration Argentometric:≥98.5 % (on dried substance)
- Loss on drying:≤1 %
- Iodate (IO3):≤4 ppm
- Sulfate (SO4):≤200 ppm
- Thiosulphate (S2O3):≤100 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤10 ppm