Order Entry
Germany
ContactUsLinkComponent
Potassium dichromate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Oxidizing
Toxic
Corrosive
Health
Environment
Potassium dichromate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Oxidizing
Toxic
Corrosive
Health
Environment
Potassium dichromate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    130 g/l (20 °C)
  • Melting Point:
    398 °C
  • Molar Mass:
    294.19 g/mol
  • Bulk Density:
    1250 kg/m3
  • Boiling Point:
    500 °C (1013 hPa)
  • Density:
    2.69 g/cm3 (20 °C)
  • pH:
    3.6 (100 g/l, H2O)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information