Order Entry
Germany
ContactUsLinkComponent
 
Sodium chloride ≥99.5%, Gen-Apex for molecular biology
Catalog # 33614.265
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chloride ≥99.5%, Gen-Apex for molecular biology
Catalog # 33614.265
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Colouration (1 mol/l; water):
    Max. 10 APHA
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Transmittance (230 nm):
    Min. 90 %
  • Transmittance (260 nm):
    Min. 95 %
  • Transmittance (280 nm):
    Min. 97 %
  • Transmittance (320 nm):
    Min. 99 %

Specification Test Results

About this item

 :
 :58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477