Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
Specification Test Results
- Assay:Min. 98 %
- Appearance:Colourless crystals
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 33L Plastic bucket UN Z (ZPACK13247037) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
spec_VWRC27788.297 spec_VWRC27788.297 spec_VWRC27788.366 spec_VWRC27788.366 spec_VWRC27788.460
Product specification
More
VWR Chemicals Big Packs and Bulks