Sodium chloride ≥99.0%, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
EINECS:231-598-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (NaCl):≥99.0 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- pH of 5% Solution at 25°C:5.5 - 7.5
- Insoluble Matter:≤0.005 %
- Bromide (Br):≤0.01 %
- Chlorate and Nitrate (as NO₃):≤0.003 %
- Iodide (I):≤0.002 %
- ACS - Sulfate (SO₄):≤0.003 %
- Barium (Ba):≤0.001 %
- Calcium, Magnesium, and R₂O₃ Precipitate:≤0.005 %
- Potassium (K):≤0.005 %
- Nitrogen Compounds (as N):≤5 ppm
- Phosphate (PO₄):≤5 ppm
- Heavy Metals (as Pb):≤2 ppm
- Trace Impurities - Iron (Fe):≤1 ppm