Lithium chloride ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
:42.39 g/mol
Merck Index:13,05550
Boiling Pt:1382 °C (1013 hPa)
Melting Pt:614 °C
Density:2.07 g/cm³ (20 °C)
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99.0 %
- pH (20°C; 5 %):5 - 9
- Heavy metals (as Pb):Max. 5 ppm
- Total N (Nitrogen):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ba (Barium):Max. 20 ppm
- Ca (Calcium):Max. 50 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 0.05 %
- Mg (Magnesium):Max. 50 ppm
- Na (Sodium):Max. 0.05 %
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
25012_LITHIUM CHLORIDE ANALAR NORMAPUR REAG.PE_BSE_REV0.pdf
BSE/TSE Certificates
spec_VWRC25012.237 spec_VWRC25012.260 spec_VWRC25012.260 spec_VWRC25012.363 spec_VWRC25012.363
Product specification
More