Lithium chloride
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
:42.39 g/mol
Merck Index:13,05550
Boiling Pt:1382 °C (1013 hPa)
Melting Pt:614 °C
Density:2.07 g/cm³ (20 °C)
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient