Cesium chloride, Supelco®
Catalog # 1.02041.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Solubility:1860 g/l (20 °C)
- Melting Point:646 °C
- Molar Mass:168.36 g/mol
- Bulk Density:1800 kg/m3
- Boiling Point:1382 °C
- Density:3.97 g/cm3 (20 °C)
- pH:7 - 9 (50 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:168.36 g/mol
Merck Index:13,02021
Boiling Pt:1303 °C (1013 hPa)
Melting Pt:646 °C
Density:3.7808 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-600-2
:GHS08
CAS Number:7647-17-8
Formula:CsCl
MDL Number:MFCD00010955