Order Entry
Germany
ContactUsLinkComponent
Cesium chloride ≥99.5%, Puriss. p.a., Fluka™
Cesium chloride ≥99.5%, Puriss. p.a., Fluka™
Certificates
Cesium chloride ≥99.5%, Puriss. p.a., Fluka™

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Barium (Ba):
    Max. 0.002 %
  • Calcium (Ca):
    Max. 0.002 %
  • Copper (Cu):
    Max. 0.0002 %
  • Iron (Fe):
    Max. 0.0005 %
  • Potassium (K):
    Max. 0.002 %
  • Magnesium (Mg):
    Max. 0.0005 %
  • Sodium (Na):
    Max. 0.002 %
  • Lead (Pb):
    Max. 0.0001 %
  • Rubidium (Rb):
    Max. 0.005 %
  • Zinc (Zn):
    Max. 0.0002 %
  • Sulfate (SO₄):
    Max. 0.005 %
  • Total N:
    Max. 0.001 %

Specification Test Results

Related Information