Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cesium chloride
Cesium chloride ≥99.9% (metals basis)
Cesium chloride ≥99.9% (metals basis)
:
Thermo Fisher Scientific
Cesium chloride ≥99.9% (metals basis)
:
Thermo Fisher Scientific
:
7647-17-8
:
:
CsCl
:
MFCD00010955
:
168.36 g/mol
:
14,2011
:
1303 °C
:
646 °C
:
3.99
:
Ambient
:
231-600-2
msds_ALFA010018
Thermo Scientific Acroseal 25 Years