Sodium hydrogen carbonate
Catalog # PROONIST-2191A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:30 g
- Seq:3332
Specifications
About this item
:
:84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528