Order Entry
Germany
ContactUsLinkComponent
Sodium hydrogen carbonate FCC, E500, SAFC®
Sodium hydrogen carbonate FCC, E500, SAFC®
Catalog # 1.72577.9025
Supplier:  Merck
CAS Number:  
undefined
Sodium hydrogen carbonate FCC, E500, SAFC®
Catalog # 1.72577.9025
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528