Sodium hydrogen carbonate, BAKER, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-633-8
Specification Test Results
- Assay:99.0 - 101.0 %
- Ammonium (NH4):max. 20 ppm
- Appearance of solution:passes test
- Carbonates:passes test
- Arsenic (As):max. 2 ppm
- Calcium (Ca):max. 100 ppm
- Chloride (Cl):max. 150 ppm
- Iron (Fe):max. 20 ppm
- Heavy Metals (as Pb):max. 10 ppm
- Identification:passes test
- Sulfates (as SO4):max. 150 ppm