Sodium hydrogen carbonate 99.7-100.3% ACS
:144-55-8
:
:NaHCO₃
:MFCD00003528
:84.01 g/mol
:13,08655
:270 °C
:2.159 g/cm³ (20 °C)
:Ambient
:205-633-8
- Ammonium:<0.0005%
- Calcium:≤0.02%
- Chloride:≤0.003%
- Heavy metals (as Pb):≤0.0005%
- Insolubles:≤0.015%
- Iron:≤0.001%
- Magnesium:≤0.005%
- Phosphate:≤0.001%
- Potassium:≤0.005%
- Purity:99.7 - 100.3%
- Sulphur compounds:≤0.003%