Sodium hydrogen carbonate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Merck Index:14,8583
Melting Pt:270 °C
Density:2.159
Storage Temperature:Ambient
EINECS:205-633-8