Order Entry
Germany
ContactUsLinkComponent
Sodium carbonate ≥97%, TECHNICAL
Irritant
Sodium carbonate ≥97%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Sodium carbonate ≥97%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 97 %

Specification Test Results