Potassium carbonate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:584-08-7
:
Formula:K₂CO₃
MDL Number:MFCD00011382
:138.21 g/mol
Merck Index:13,07702
Melting Pt:891 °C
Density:2.42 g/cm³ (20 °C)
EINECS:209-529-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (on anhydrous substance):Min. 99.0 %
- Ba (Barium):Passes test
- CN (Cyanide):Passes test
- Sulphur compounds (as S):Passes test
- Free potassium hydroxide:Max. 0.05 %
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Substances precipitated by NH4OH:Max. 100 ppm
- Cl (Chloride):Max. 30 ppm
- NO3 (Nitrate):Max. 30 ppm
- PO4 (Phosphate):Max. 10 ppm
- SO4 + SO3 (as SO4):Max. 20 ppm
- As (Arsenic):Max. 2 ppm
- Ca + Mg (as Ca):Max. 100 ppm
- Fe (Iron):Max. 5 ppm
- Na (Sodium):Max. 0.2 %
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 50L Plastic bucket UN Y (ZPACK13247050) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
26726_POTASSIUM CARBONATE ANALAR NP REAG.PE_BSE_REV0.pdf
BSE/TSE Certificates
spec_VWRC26726.260 spec_VWRC26726.260 spec_VWRC26726.297 spec_VWRC26726.297 spec_VWRC26726.297
Product specification
More