Order Entry
Germany
ContactUsLinkComponent
 
Potassium carbonate ≥98%, GPR RECTAPUR®
 
undefined
Potassium carbonate ≥98%, GPR RECTAPUR®
 

 

  • Assay (on anhydrous substance):
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 + SO3 (as SO4):
    Max. 100 ppm
  • Fe (Iron):
    Max. 20 ppm