Potassium cyanide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:151-50-8
:
Formula:KCN
MDL Number:MFCD00011397
:65.12 g/mol
Merck Index:14,7626
Boiling Pt:1625 °C
Melting Pt:634 °C
Density:1.52
Storage Temperature:Ambient
EINECS:205-792-3
UN:1680
ADR:6.1,I
:GHS06
:GHS08
:GHS09