Sodium bromide, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-15-6
:
Formula:NaBr
MDL Number:MFCD00003475
:102.89 g/mol
Merck Index:14,8594
Boiling Pt:1390 °C
Melting Pt:755 °C
Density:3.203
:ampouled under argon
Storage Temperature:Ambient
EINECS:231-599-9