0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Sodium bromide
Sodium bromide ≥99.0% ACS
Sodium bromide ≥99.0% ACS
:
Thermo Fisher Scientific
Sodium bromide ≥99.0% ACS
:
Thermo Fisher Scientific
:
7647-15-6
:
:
NaBr
:
MFCD00003475
:
102.89 g/mol
:
14,8594
:
1390 °C
:
755 °C
:
3.203
:
Ambient
:
231-599-9
ALFA36444.A3_TDS_EN
externalurl.pdf
msds_ALFA036444
Thermo Scientific Acroseal 25 Years