Centrifuge tubes, Axygen®
Supplier: Corning
About this item
PP, conical, printed graduation, with HDPE flat screw caps.
- Max. RCF: 12 000×g
- Easy one hand operation
- Large writing area for sample identification
6 Options Available Below
Product Details & Documents
PP, conical, printed graduation, with HDPE flat screw caps.
- Max. RCF: 12 000×g
- Easy one hand operation
- Large writing area for sample identification
Specifications for Product Family
Specifications
- Catalog No:
- 525-0518
- 525-0780
- 525-1746
- 525-0519
- 525-0781
- 525-1747
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Product Information
- CapacityClosure typeVersionPackedAvailabilityPrice
- 15 mlScrew capNon-sterile500 tubes and 500 unassembled caps per caseLog in for availability1 * 500 ST165,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.525-0518Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.525-0780Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - 15 mlScrew capSterile25 tubes and assembled caps per rackLog in for availability1 * 500 ST242,00 €Specifications:
More
Specifications
Cat. No.525-1746Environmentally PreferableEnvironmental certification - 50 mlScrew capSterile500 tubes and 500 unassembled caps per caseLog in for availability1 * 500 ST152,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.525-0519Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.525-0781Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - 50 mlScrew capSterile25 tubes and assembled caps per rack, 20 racks per caseLog in for availability1 * 500 ST203,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.525-1747Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact