1,4-Dioxane, dehydrated (max. 0.01% H₂O) ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Catalog # 23539.291
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Acidity or alkalinity:Max. 0.001 meq/g
- Colouration:Max. 10 APHA
- Peroxides (as H2O2):Max. 0.0100 %
- Water:Max. 0.0100 %
- Acetyls and carbonyl derivatives:Max. 40 ppm
- Evaporation residue:Max. 50 ppm
- Fe (Iron):Max. 1 ppm
Specification Test Results
About this item
:
:88.11 g/mol
Merck Index:13,03330
Boiling Pt:101.1 °C (1013 hPa)
Melting Pt:11.8 °C
Density:1.03 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:204-661-8
UN:1165
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 20 ppm
:GHS02
:GHS08
:GHS07
CAS Number:123-91-1
Formula:C₄H₈O₂
MDL Number:MFCD00006571