1,4-Dioxane stabilised, EMPLURA®, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Merck Index:13,03330
Boiling Pt:101.1 °C (1013 hPa)
Melting Pt:11.8 °C
Density:1.03 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:12 °C
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 25 ppm
EINECS:204-661-8
UN:1165
:GHS02
:GHS08
:GHS07
Related Information
MERC1.03115.2500_TDS_EN ProductPage_103115 ProductPage_103115
Product specification
msds_103115