Order Entry
Germany
ContactUsLinkComponent
1,5-Diazobicyclo[4.3.0]-5-nonene for synthesis, Sigma-Aldrich®
Corrosive
1,5-Diazobicyclo[4.3.0]-5-nonene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
1,5-Diazobicyclo[4.3.0]-5-nonene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    124.18 g/mol
  • Boiling Point:
    110 °C (20 hPa)
  • Flash Point:
    94 °C
  • Refractive Index:
    1.519 (20 °C, 589 nm)
  • Density:
    1.04 g/cm3 (20 °C)

Specification Test Results

Related Information