
1-Methylcyclopentene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:693-89-0
:
Formula:C₆H₁₀
MDL Number:MFCD00001397
:82.15 g/mol
Boiling Pt:73…75 °C
Melting Pt:–142 °C
Density:0.779
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:211-762-0
UN:3295
ADR:3,II
:GHS02