Cyclopentane, Supelco®
Supplier: Merck
Jump to:
About this item
Synonyms: Pentamethylene
- CAS Number:287-92-3
- Formula:C₅H₁₀
- MDL Number:MFCD00001356
- Molecular Weight:70.13 g/mol
- Merck Index:13,02769
- Boiling Pt:49 °C (1013 hPa)
- Melting Pt:–93.3 °C
- Density:0.746 g/cm³ (20 °C)
- Flash Pt:–41 °C
- Storage Temperature:Ambient
- UN:1146
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:206-016-6
2 Options Available Below
Product Details & Documents
Synonyms: Pentamethylene
- CAS Number:287-92-3
- Formula:C₅H₁₀
- MDL Number:MFCD00001356
- Molecular Weight:70.13 g/mol
- Merck Index:13,02769
- Boiling Pt:49 °C (1013 hPa)
- Melting Pt:–93.3 °C
- Density:0.746 g/cm³ (20 °C)
- Flash Pt:–41 °C
- Storage Temperature:Ambient
- UN:1146
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:206-016-6



