Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Supelco® Organic Reference Standard, Benzene-d₆ solution

Benzene-d₆ solution, 2000 μg/ml in methanol, Certified reference material, 1
Catalog # 48940-U
Supplier:   Merck
Supelco® Organic Reference Standard, Benzene-d₆ solution
Catalog # 48940-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Benzene-d₆ solution

Benzene-d₆ solution, 2000 μg/ml in methanol, Certified reference material, 1

CAS Number:  
Price
37,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
  • GHS signal: GHS06, GHS08

 : P201 - P210 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311

Specifications

Specifications

  • Concentration:
    2000 μg/ml in methanol
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Benzene-d₆ solution
  • Cat. No.:
    48940-U
  • M-Clarity quality level:
    MQ100
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H340 - H350 - H370
  • Molecular weight:
    84,15
  • CAS No.:
    1076-43-3
  • Shipping conditions:
    STD

Specifications