1,1'-Bis(diphenylphosphino)ferrocene ≥99%
Catalog # STRE26-0270-1G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:554.39 g/mol
Melting Pt:180 °C
EINECS:430-420-3
:GHS07
CAS Number:12150-46-8
Formula:C₃₄H₂₈FeP₂
MDL Number:MFCD00001422