Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DL-Tropic acid
DL-Tropic acid, Sigma-Aldrich®
DL-Tropic acid, Sigma-Aldrich®
SIALT89206-25G
:
Merck
:
DL-Tropic acid, Sigma-Aldrich®
SIALT89206-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
166.18 g/mol
:
M-Clarity™ quality level = MQ100
:
208-465-3
:
529-64-6
:
552-63-6
:
C₉H₁₀O₃
:
MFCD00004255