Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2-furoate
Ethyl-2-furoate, Sigma-Aldrich®
Ethyl-2-furoate, Sigma-Aldrich®
SIALE28501-100G
:
Merck
:
Ethyl-2-furoate, Sigma-Aldrich®
SIALE28501-100G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
140.14 g/mol
:
M-Clarity™ quality level = MQ100
:
210-404-0
:
614-99-3
:
C₇H₈O₃
:
MFCD00003237