Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α-Acetyl-γ-butyrolactone
α-Acetyl-γ-butyrolactone, Sigma-Aldrich®
α-Acetyl-γ-butyrolactone, Sigma-Aldrich®
α-Acetyl-γ-butyrolactone, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
α-Acetyl-γ-butyrolactone, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
517-23-7
:
Formula:
C₆H₈O₃
MDL Number:
MFCD00005394
:
128.13 g/mol
EINECS:
208-235-2